siRNA Details
| Gene Symbol: Mpnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026530.5 | NP_080806.4 | 6 | 776 | ||
| GenBank mRNA | AK012128.1 | - | 5 | 731 | ||
| AK146973.1 | - | 6 | 761 | |||
| AK160356.1 | - | 6 | 745 | |||
| AK172381.1 | - | 5 | 519 | |||
| AK213160.1 | - | 4 | 265 | |||
| BC017625.1 | - | 5 | 725 | |||

Mpnd
Genomic Map