siRNA Details
Gene Symbol: Aen | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162939.1 | NP_001156411.1 | 2, 3 | 569 | ||
NM_026531.4 | NP_080807.3 | 2, 3 | 683 | |||
GenBank mRNA | AK012555.1 | - | 2, 3 | 668 | ||
AK031972.1 | - | 2, 3 | 684 | |||
AK089090.1 | - | 2, 3 | 505 | |||
AK090056.1 | - | 2, 3 | 552 | |||
AK165708.1 | - | 2, 3 | 563 | |||
AK166806.1 | - | 2, 3 | 546 | |||
AK167042.1 | - | 2, 3 | 546 | |||
AK167658.1 | - | 2, 3 | 642 | |||
BC022614.1 | - | 2, 3 | 763 | 4167800 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
