siRNA Details
| Gene Symbol: Dusp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024438.4 | NP_077758.1 | 1 | 199 | ||
| GenBank mRNA | AB038769.1 | - | 1 | 154 | ||
| AF237618.1 | - | 1 | 371 | |||
| AK010127.1 | - | 1 | 196 | |||
| AK049927.1 | - | 1 | 164 | |||
| AK077871.1 | - | 1 | 200 | |||
| AK083610.1 | - | 148 | ||||
| BC021591.1 | - | 1 | 154 | 5149051 | ||

Dusp19
Genomic Map