siRNA Details
| Gene Symbol: Dusp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024438.4 | NP_077758.1 | 1 | 284 | ||
| GenBank mRNA | AB038769.1 | - | 1 | 239 | ||
| AB051896.1 | - | 1 | 258 | |||
| AF237618.1 | - | 1 | 456 | |||
| AK010127.1 | - | 1 | 281 | |||
| AK049927.1 | - | 1 | 249 | |||
| AK077871.1 | - | 1 | 285 | |||
| AK083610.1 | - | 233 | ||||
| BC021591.1 | - | 1 | 239 | 5149051 | ||

Dusp19
Genomic Map