siRNA Details
| Gene Symbol: Snhg17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040893.1 | - | 3 | 1452 | ||
| AK046044.1 | - | 4 | 535 | |||
| AK079109.1 | - | 3 | 1461 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snhg17| Gene Symbol: Snhg17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040893.1 | - | 3 | 1452 | ||
| AK046044.1 | - | 4 | 535 | |||
| AK079109.1 | - | 3 | 1461 | |||