siRNA Details
| Gene Symbol: Sdccag3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085407.1 | NP_001078876.1 | 4 | 565 | ||
| NM_001085408.1 | NP_001078877.1 | 3 | 496 | |||
| NM_026563.3 | NP_080839.2 | 5 | 715 | |||
| GenBank mRNA | AK014885.1 | - | 5 | 684 | ||
| AK021273.1 | - | 5 | 706 | |||
| AK082787.1 | - | 4 | 536 | |||
| AK163690.1 | - | 3 | 467 | |||
| BC005612.1 | - | 3 | 487 | |||
| BC031199.1 | - | 3 | 445 | 5064948 | ||
| BC061037.1 | - | 3 | 776 | |||

Sdccag3
Genomic Map