siRNA Details
| Gene Symbol: B230217C12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080935.1 | NP_001074404.1 | 1 | 59 | ||
| GenBank mRNA | AK081899.1 | - | 14 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B230217C12Rik| Gene Symbol: B230217C12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080935.1 | NP_001074404.1 | 1 | 59 | ||
| GenBank mRNA | AK081899.1 | - | 14 | |||