siRNA Details
| Gene Symbol: Upf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026573.2 | NP_080849.1 | 3 | 381 | ||
| GenBank mRNA | AK011606.1 | - | 3 | 379 | ||
| AK019989.1 | - | 3 | 359 | |||
| AK145431.1 | - | 3 | 384 | |||
| AK162222.1 | - | 3 | 377 | |||
| BC047049.1 | - | 1 | 40 | |||
| BC058761.1 | - | 3 | 362 | |||
| BC117769.1 | - | 3 | 399 | 40091025 | ||
| BC117770.1 | - | 3 | 399 | 40091027 | ||

Upf3b
Genomic Map