siRNA Details
| Gene Symbol: Ndnf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172399.3 | NP_765987.2 | 4 | 2158 | ||
| GenBank mRNA | AK017759.1 | - | 1 | 70 | ||
| AK029210.1 | - | 4 | 2159 | |||
| AK049802.1 | - | 4 | 2183 | |||
| BC047154.1 | - | 4 | 2132 | 6489681 | ||

Ndnf
Genomic Map