siRNA Details
| Gene Symbol: D730048I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026593.3 | NP_080869.1 | 3 | 296 | ||
| GenBank mRNA | AK020299.1 | - | 3 | 268 | ||
| AK020302.1 | - | 3 | 268 | |||
| AK020310.1 | - | 3 | 249 | |||
| AK021351.1 | - | 3 | 268 | |||
| AK033813.1 | - | 3 | 297 | |||
| BC027498.1 | - | 3 | 230 | 3465762 | ||

D730048I06Rik
Genomic Map