siRNA Details
| Gene Symbol: Leprotl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026609.2 | NP_080885.1 | 3 | 191 | ||
| GenBank mRNA | AK003329.1 | - | 3 | 202 | ||
| AK013870.1 | - | 3 | 261 | |||
| AK075740.1 | - | 3 | 246 | |||
| AK150289.1 | - | 3 | 250 | |||
| AK153277.1 | - | 3 | 236 | |||
| AK154712.1 | - | 3 | 256 | |||
| AK159401.1 | - | 3 | 179 | |||
| BC004677.1 | - | 3 | 191 | 3500223 | ||

Leprotl1
Genomic Map