siRNA Details
| Gene Symbol: Leprotl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026609.2 | NP_080885.1 | 1, 2 | 53 | ||
| GenBank mRNA | AK003329.1 | - | 1, 2 | 64 | ||
| AK013870.1 | - | 1, 2 | 123 | |||
| AK075740.1 | - | 1, 2 | 108 | |||
| AK150289.1 | - | 1, 2 | 112 | |||
| AK153277.1 | - | 1, 2 | 98 | |||
| AK154712.1 | - | 1, 2 | 118 | |||
| AK159401.1 | - | 1, 2 | 41 | |||
| BC004677.1 | - | 1, 2 | 53 | 3500223 | ||

Leprotl1
Genomic Map