siRNA Details
| Gene Symbol: Ndufb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026612.3 | NP_080888.1 | 1, 2 | 140 | ||
| GenBank mRNA | AK007495.1 | - | 1, 2 | 116 | ||
| AK010642.1 | - | 1, 2 | 147 | |||
| AK019008.1 | - | 1, 2 | 145 | |||
| AK028334.1 | - | 149 | ||||
| BC013510.1 | - | 1, 2 | 150 | 4225177 | ||

Ndufb2
Genomic Map