siRNA Details
| Gene Symbol: Urm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026615.4 | NP_080891.1 | 5 | 265 | ||
| GenBank mRNA | AK012124.1 | - | 5 | 271 | ||
| AK019352.1 | - | 5 | 271 | |||
| AK049227.1 | - | 5 | 266 | |||
| AK049849.1 | - | 5 | 266 | |||
| AK165008.1 | - | 2 | 1265 | |||
| AK187514.1 | - | 2 | 64 | |||
| AK210360.1 | - | 6 | 331 | |||
| AK215344.1 | - | 2 | 64 | |||
| BC026994.1 | - | 5 | 257 | 5368627 | ||
| BC060255.1 | - | 2 | 1138 | |||

Urm1
Genomic Map