siRNA Details
| Gene Symbol: Rnaseh2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026616.2 | NP_080892.1 | 3 | 416 | ||
| GenBank mRNA | AF346604.1 | - | 3 | 414 | ||
| AK011556.1 | - | 3 | 394 | |||
| AK018776.1 | - | 3 | 405 | |||
| BC024333.1 | - | 3 | 388 | 5040687 | ||

Rnaseh2c
Genomic Map