siRNA Details
| Gene Symbol: Nudt21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026623.3 | NP_080899.1 | 1 | 247 | ||
| GenBank mRNA | AK011688.2 | - | 1 | 237 | ||
| AK012308.1 | - | 1 | 221 | |||
| AK019433.1 | - | 1 | 230 | |||
| AK020150.1 | - | 1 | 227 | |||
| AK146419.1 | - | 1 | 213 | |||
| AK147061.1 | - | 230 | ||||
| AK160147.1 | - | 1 | 213 | |||
| AK204985.1 | - | 1 | 127 | |||
| BC008270.1 | - | 1 | 230 | 2811199 | ||
| BC090834.1 | - | 206 | 6855233 | |||

Nudt21
Genomic Map