siRNA Details
| Gene Symbol: Nudt21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026623.3 | NP_080899.1 | 6, 7 | 803 | ||
| GenBank mRNA | AK011688.2 | - | 6, 7 | 793 | ||
| AK012308.1 | - | 6, 7 | 777 | |||
| AK019433.1 | - | 6, 7 | 786 | |||
| AK020150.1 | - | 6, 7 | 784 | |||
| AK146419.1 | - | 6, 7 | 769 | |||
| AK147061.1 | - | 786 | ||||
| AK160147.1 | - | 6, 7 | 769 | |||
| AK213941.1 | - | 1, 2 | 57 | |||
| BC008270.1 | - | 6, 7 | 786 | 2811199 | ||
| BC090834.1 | - | 762 | 6855233 | |||

Nudt21
Genomic Map