siRNA Details
| Gene Symbol: Wfdc15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183271.2 | NP_899094.1 | 1 | 142 | ||
| GenBank mRNA | AK006775.1 | - | 1 | 136 | ||
| AK018854.1 | - | 1 | 136 | |||
| AK018888.1 | - | 1 | 143 | |||
| AK018909.1 | - | 1 | 114 | |||
| BC100423.1 | - | 1 | 121 | 30919644 | ||

Wfdc15a
Genomic Map