siRNA Details
Gene Symbol: 1700102H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015782.1 | - | 3 | 223 | ||
AK018936.1 | - | 2 | 106 | |||
AK018990.1 | - | 2 | 106 | |||
BY715326.1 | - | 3 | 223 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700102H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015782.1 | - | 3 | 223 | ||
AK018936.1 | - | 2 | 106 | |||
AK018990.1 | - | 2 | 106 | |||
BY715326.1 | - | 3 | 223 |