siRNA Details
| Gene Symbol: Fam195a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026633.3 | NP_080909.1 | 2 | 146 | ||
| GenBank mRNA | AK003564.1 | - | 2 | 148 | ||
| AK020614.1 | - | 2 | 145 | |||
| BC024332.1 | - | 2 | 138 | 5011430 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam195a| Gene Symbol: Fam195a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026633.3 | NP_080909.1 | 2 | 146 | ||
| GenBank mRNA | AK003564.1 | - | 2 | 148 | ||
| AK020614.1 | - | 2 | 145 | |||
| BC024332.1 | - | 2 | 138 | 5011430 | ||