siRNA Details
| Gene Symbol: Zdhhc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026647.3 | NP_080923.2 | 4 | 555 | ||
| GenBank mRNA | AK144108.1 | - | 4 | 454 | ||
| AK148344.1 | - | 4 | 448 | |||
| BC043110.1 | - | 4 | 401 | 6406930 | ||
| BC058372.1 | - | 4 | 423 | |||

Zdhhc21
Genomic Map