siRNA Details
| Gene Symbol: Zdhhc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026647.3 | NP_080923.2 | 7, 8 | 984 | ||
| GenBank mRNA | AK020293.1 | - | 7, 8 | 866 | ||
| AK144108.1 | - | 6, 7 | 745 | |||
| AY166674.1 | - | 5, 6 | 622 | |||
| BC043110.1 | - | 7, 8 | 830 | 6406930 | ||
| BC058372.1 | - | 7, 8 | 852 | |||

Zdhhc21
Genomic Map