siRNA Details
| Gene Symbol: Ddx39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197982.3 | NP_932099.2 | 10 | 1418 | ||
| GenBank mRNA | AK011285.1 | - | 9 | 1257 | ||
| AK012002.1 | - | 10 | 1325 | |||
| AK016960.1 | - | 10 | 1327 | |||
| AK088894.1 | - | 4 | 720 | |||
| AK137114.1 | - | 1324 | ||||
| AK145927.1 | - | 10 | 1361 | |||
| AK146294.1 | - | 10 | 1363 | |||
| AK149475.1 | - | 10 | 1760 | |||
| AK150997.1 | - | 10 | 1347 | |||
| AK151395.1 | - | 11 | 1387 | |||
| AK152394.1 | - | 10 | 1328 | |||
| AK162752.1 | - | 1886 | ||||
| AK164289.1 | - | 10 | 1288 | |||
| AK167890.1 | - | 10 | 1288 | |||
| AK168079.1 | - | 10 | 1287 | |||
| AK172448.1 | - | 10 | 1347 | |||
| BC020134.1 | - | 10 | 1306 | 3969048 | ||

Ddx39
Genomic Map