siRNA Details
| Gene Symbol: Ncapd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146171.1 | NP_666283.1 | 24 | 3109 | ||
| GenBank mRNA | AK019259.2 | - | 10 | 1273 | ||
| AK129068.1 | - | 24 | 3074 | |||
| AK156143.1 | - | 24 | 3140 | |||
| BC006673.1 | - | 17 | 2255 | |||
| BC008592.1 | - | 15 | 2003 | |||
| BC023750.1 | - | 10 | 1138 | |||
| BC025460.1 | - | 24 | 3129 | |||
| BC026982.1 | - | 24 | 3109 | 5364933 | ||
| BC029133.1 | - | 24 | 3066 | 4951596 | ||

Ncapd2
Genomic Map