siRNA Details
| Gene Symbol: Tmem174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026685.2 | NP_080961.1 | 1 | 123 | ||
| GenBank mRNA | AK002365.1 | - | 1 | 50 | ||
| AK057630.1 | - | 1 | 51 | |||
| BC030878.1 | - | 1 | 72 | 4241631 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem174| Gene Symbol: Tmem174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026685.2 | NP_080961.1 | 1 | 123 | ||
| GenBank mRNA | AK002365.1 | - | 1 | 50 | ||
| AK057630.1 | - | 1 | 51 | |||
| BC030878.1 | - | 1 | 72 | 4241631 | ||