siRNA Details
| Gene Symbol: 0610030E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026696.1 | NP_080972.1 | 3 | 447 | ||
| GenBank mRNA | AK002701.1 | - | 3 | 393 | ||
| AK085662.1 | - | 1 | 1342 | |||
| BC064080.1 | - | 3 | 430 | 1246433 | ||
| BC066053.1 | - | 3 | 421 | 6827710 | ||
| BC117811.1 | - | 2 | 474 | |||
| BC117812.1 | - | 3 | 397 | 40092656 | ||

0610030E20Rik
Genomic Map