siRNA Details
Gene Symbol: Tmem129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026698.2 | NP_080974.2 | 2 | 430 | ||
GenBank mRNA | AK002836.1 | - | 2 | 479 | ||
AK172044.1 | - | 2 | 559 | |||
BC029088.1 | - | 2 | 476 | 3587662 | ||
BC094042.1 | - | 2 | 430 | 5098723 |
