siRNA Details
| Gene Symbol: Ankrd13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026720.2 | NP_080996.2 | 11 | 1296 | ||
| GenBank mRNA | AK089186.1 | - | 11 | 1289 | ||
| BC058982.1 | - | 11 | 1207 | 5685059 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd13d| Gene Symbol: Ankrd13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026720.2 | NP_080996.2 | 11 | 1296 | ||
| GenBank mRNA | AK089186.1 | - | 11 | 1289 | ||
| BC058982.1 | - | 11 | 1207 | 5685059 | ||