siRNA Details
| Gene Symbol: Slc39a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290765.1 | NP_001277694.1 | 4 | 708 | ||
| NM_026721.3 | NP_080997.1 | 4 | 669 | |||
| GenBank mRNA | AK003191.1 | - | 1 | 73 | ||
| AK035296.1 | - | 4 | 576 | |||
| AK156951.1 | - | 4 | 594 | |||
| BC100463.1 | - | 4 | 676 | 30918840 | ||
| BC125525.1 | - | 4 | 530 | 40130040 | ||
| BC137752.1 | - | 4 | 530 | 8860771 | ||

Slc39a13
Genomic Map