siRNA Details
| Gene Symbol: Fbxl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133694.2 | NP_598455.2 | 4 | 1178 | ||
| GenBank mRNA | AK003032.1 | - | 4 | 901 | ||
| AK145009.1 | - | 4 | 1138 | |||
| AK172339.1 | - | 5 | 1075 | |||
| AW124512.1 | - | 1 | 184 | |||
| Gene Symbol: Cuedc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164290.1 | NP_001157762.1 | 10 | 1473 | ||
| NM_001164291.1 | NP_001157763.1 | 9 | 1365 | |||
| NM_001164292.1 | NP_001157764.1 | 9 | 1362 | |||
| NM_001164293.1 | NP_001157765.1 | 9 | 1332 | |||
| NM_001164294.1 | NP_001157766.1 | 9 | 1311 | |||
| NM_001164295.1 | NP_001157767.1 | 9 | 1308 | |||
| NM_024192.2 | NP_077154.2 | 10 | 1464 | |||
| GenBank mRNA | AK013884.1 | - | 9 | 1323 | ||

Fbxl15
Genomic Map