siRNA Details
| Gene Symbol: 1010001N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003136.1 | - | 2 | 177 | ||
| AK033147.1 | - | 1 | 204 | |||
| CK332479.1 | - | 139 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1010001N08Rik| Gene Symbol: 1010001N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003136.1 | - | 2 | 177 | ||
| AK033147.1 | - | 1 | 204 | |||
| CK332479.1 | - | 139 | ||||