siRNA Details
| Gene Symbol: Mrpl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026732.2 | NP_081008.1 | 3 | 490 | ||
| GenBank mRNA | AK003500.1 | - | 3 | 457 | ||
| AK050273.1 | - | 3 | 476 | |||
| AK144153.1 | - | 3 | 494 | |||
| BC027021.1 | - | 3 | 490 | 5374870 | ||

Mrpl14
Genomic Map