siRNA Details
| Gene Symbol: 1110007C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026738.2 | NP_081014.1 | 3 | 278 | ||
| GenBank mRNA | AK003526.1 | - | 3 | 279 | ||
| AK154517.1 | - | 3 | 250 | |||
| BC023485.1 | - | 4 | 410 | |||
| BC047205.1 | - | 3 | 318 | |||
| BC147073.1 | - | 3 | 234 | 9056051 | ||
| BC147074.1 | - | 3 | 234 | 9056042 | ||

1110007C09Rik
Genomic Map