siRNA Details
| Gene Symbol: 1110007C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026738.2 | NP_081014.1 | 6 | 569 | ||
| GenBank mRNA | AK003526.1 | - | 6 | 570 | ||
| AK154517.1 | - | 6 | 541 | |||
| BC023485.1 | - | 7 | 701 | |||
| BC038930.1 | - | 2 | 449 | |||
| BC047205.1 | - | 6 | 609 | |||
| BC147073.1 | - | 6 | 525 | 9056051 | ||
| BC147074.1 | - | 6 | 525 | 9056042 | ||

1110007C09Rik
Genomic Map