siRNA Details
| Gene Symbol: Zfp579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026741.2 | NP_081017.1 | 2 | 748 | ||
| GenBank mRNA | AK143550.1 | - | 1 | 1804 | ||
| AK148159.1 | - | 1 | 1845 | |||
| BC048385.1 | - | 1 | 1078 | 6309639 | ||
| BC054068.1 | - | 1 | 1078 | |||

Zfp579
Genomic Map