siRNA Details
| Gene Symbol: Ptx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163416.1 | NP_001156888.1 | 3 | 1110 | ||
| NM_026747.1 | NP_081023.1 | 3 | 1200 | |||
| GenBank mRNA | AK003777.1 | - | 2 | 669 | ||
| BC107380.2 | - | 3 | 1093 | 40057084 | ||
| BC107381.2 | - | 3 | 1183 | 40057088 | ||
| BC115902.1 | - | 3 | 738 | |||
| BC118508.1 | - | 3 | 1183 | 40106397 | ||

Ptx4
Genomic Map