siRNA Details
| Gene Symbol: Ints1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026748.2 | NP_081024.3 | 31 | 4666 | ||
| GenBank mRNA | AB257854.1 | - | 31 | 4626 | ||
| AK148183.1 | - | 2 | 124 | |||
| BC010333.1 | - | 2 | 247 | |||
| BC063266.1 | - | 30 | 4434 | 6409930 | ||

Ints1
Genomic Map