siRNA Details
| Gene Symbol: Ints1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026748.2 | NP_081024.3 | 46 | 6730 | ||
| GenBank mRNA | AB257854.1 | - | 46 | 6690 | ||
| AK003728.1 | - | 4 | 342 | |||
| AK148183.1 | - | 17 | 2186 | |||
| BC010333.1 | - | 17 | 2311 | |||
| BC027811.1 | - | 6 | 1254 | |||
| BC051513.1 | - | 6 | 945 | |||
| BC063266.1 | - | 45 | 6498 | 6409930 | ||

Ints1
Genomic Map