siRNA Details
Gene Symbol: Eml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043335.1 | NP_001036800.1 | 4, 5 | 884 | ||
NM_001043336.2 | NP_001036801.1 | 4, 5 | 501 | |||
NM_001286346.1 | NP_001273275.1 | 4, 5 | 638 | |||
NM_001286347.1 | NP_001273276.1 | 4, 5 | 501 | |||
GenBank mRNA | AK044662.1 | - | 4, 5 | 623 | ||
AK054227.1 | - | 4, 5 | 834 | |||
BC053094.1 | - | 4, 5 | 718 | 6400458 | ||
BC059839.1 | - | 4, 5 | 479 | 6417403 | ||
BC079582.1 | - | 4, 5 | 537 | 5702743 | ||
BC098329.1 | - | 3, 4 | 280 | |||
BC125289.1 | - | 4, 5 | 472 | 40129804 | ||
BC144757.1 | - | 4, 5 | 472 | |||
BC150975.1 | - | 4, 5 | 638 | 9087888 | ||
DN172843.1 | - | 2, 3 | 268 |
