siRNA Details
| Gene Symbol: Fam189b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014995.1 | NP_001014995.1 | 4 | 969 | ||
| GenBank mRNA | BC068123.1 | - | 4 | 990 | ||
| BC085505.1 | - | 4 | 711 | |||
| BC088990.1 | - | 4 | 969 | 6402466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam189b| Gene Symbol: Fam189b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014995.1 | NP_001014995.1 | 4 | 969 | ||
| GenBank mRNA | BC068123.1 | - | 4 | 990 | ||
| BC085505.1 | - | 4 | 711 | |||
| BC088990.1 | - | 4 | 969 | 6402466 | ||