siRNA Details
| Gene Symbol: Wipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197940.2 | NP_922922.1 | 5 | 786 | ||
| GenBank mRNA | AK144539.1 | - | 4 | 1169 | ||
| BC057854.1 | - | 5 | 575 | 6494304 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wipf2| Gene Symbol: Wipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197940.2 | NP_922922.1 | 5 | 786 | ||
| GenBank mRNA | AK144539.1 | - | 4 | 1169 | ||
| BC057854.1 | - | 5 | 575 | 6494304 | ||