siRNA Details
Gene Symbol: Wipf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197940.2 | NP_922922.1 | 5 | 840 | ||
GenBank mRNA | AK144539.1 | - | 4 | 1223 | ||
BC057854.1 | - | 5 | 629 | 6494304 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wipf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197940.2 | NP_922922.1 | 5 | 840 | ||
GenBank mRNA | AK144539.1 | - | 4 | 1223 | ||
BC057854.1 | - | 5 | 629 | 6494304 |