siRNA Details
| Gene Symbol: Ecscr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033141.1 | NP_001028313.1 | 2 | 187 | ||
| GenBank mRNA | AK157485.1 | - | 2 | 187 | ||
| EU025067.1 | - | 2 | 123 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ecscr| Gene Symbol: Ecscr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033141.1 | NP_001028313.1 | 2 | 187 | ||
| GenBank mRNA | AK157485.1 | - | 2 | 187 | ||
| EU025067.1 | - | 2 | 123 | |||