siRNA Details
Gene Symbol: Ecscr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033141.1 | NP_001028313.1 | 3, 4 | 344 | ||
GenBank mRNA | AK157485.1 | - | 3, 4 | 344 | ||
EU025067.1 | - | 3, 4 | 280 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ecscr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033141.1 | NP_001028313.1 | 3, 4 | 344 | ||
GenBank mRNA | AK157485.1 | - | 3, 4 | 344 | ||
EU025067.1 | - | 3, 4 | 280 |