siRNA Details
| Gene Symbol: Ppm1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176833.4 | NP_789803.1 | 3 | 483 | ||
| GenBank mRNA | AK016942.1 | - | 3 | 484 | ||
| AK154192.1 | - | 3 | 386 | |||
| AK164964.1 | - | 3 | 364 | |||
| BC042570.1 | - | 2 | 332 | 4241448 | ||

Ppm1f
Genomic Map