siRNA Details
| Gene Symbol: Ube2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026785.2 | NP_081061.1 | 4, 5 | 483 | ||
| GenBank mRNA | AK003722.1 | - | 4, 5 | 484 | ||
| AK136186.1 | - | 4, 5 | 484 | |||
| AK160740.1 | - | 4, 5 | 515 | |||
| BC147032.1 | - | 4, 5 | 464 | 9056036 | ||
| BC147033.1 | - | 4, 5 | 464 | 9056038 | ||
| BC171983.1 | - | 4, 5 | 457 | |||

Ube2c
Genomic Map