siRNA Details
| Gene Symbol: Elac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023479.2 | NP_075968.2 | 10, 11 | 932 | ||
| GenBank mRNA | AF308696.2 | - | 10, 11 | 870 | ||
| AK156730.1 | - | 10, 11 | 933 | |||
| AK172604.1 | - | 9, 10 | 1301 | |||
| BC023134.1 | - | 10, 11 | 876 | |||
| BC048235.1 | - | 10, 11 | 866 | 6493124 | ||

Elac2
Genomic Map