siRNA Details
| Gene Symbol: Fam198b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133187.3 | NP_573450.2 | 3, 4 | 1645 | ||
| GenBank mRNA | AF285091.1 | - | 3, 4 | 1587 | ||
| AK052367.1 | - | 3, 4 | 612 | |||
| AK143522.1 | - | 3, 4 | 1376 | |||
| AK195202.1 | - | 1, 2 | 123 | |||
| BC056975.1 | - | 3, 4 | 1414 | 6827227 | ||
| EU797522.1 | - | 3, 4 | 1423 | |||

Fam198b
Genomic Map