siRNA Details
| Gene Symbol: Fam172a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163419.1 | NP_001156891.1 | 5 | 559 | ||
| NM_138312.1 | NP_612185.1 | 6 | 683 | |||
| GenBank mRNA | AF064782.1 | - | 5 | 512 | ||
| AK049727.1 | - | 5 | 520 | |||
| AK052419.1 | - | 5 | 531 | |||
| AK165273.1 | - | 6 | 582 | |||
| AK199247.1 | - | 4 | 354 | |||
| BC057380.1 | - | 5 | 527 | |||

Fam172a
Genomic Map