siRNA Details
| Gene Symbol: Fam172a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163419.1 | NP_001156891.1 | 6 | 701 | ||
| NM_001163420.1 | NP_001156892.1 | 5 | 509 | |||
| NM_138312.1 | NP_612185.1 | 7 | 825 | |||
| GenBank mRNA | AF064782.1 | - | 6 | 654 | ||
| AK165273.1 | - | 7 | 724 | |||
| AK166285.1 | - | 5 | 481 | |||
| BC057380.1 | - | 6 | 669 | |||

Fam172a
Genomic Map